Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM170A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558160
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558160 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558160 Supplier Invitrogen™ Supplier No. PA558160
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KRRQKRKHLE NEESQETAEK GGGMSKSQED ALQPGSTRVA KGWSQGVGEV TSTSEYCSCV SSSRKLIHSG IQRIHRDSPQ PQSPLAQVQE RGETPPRS Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 55%.

Acts as a nuclear transcription factor that positively regulates the expression of heat shock genes. Binds to heat shock promoter elements (HSE).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM170A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM170A
Gene Accession No. A1A519
Gene Alias FAM170A; family with sequence similarity 170 member A; family with sequence similarity 170, member A; protein FAM170A; Zinc finger domain-containing protein; zinc finger protein ZNFD; Znf domain containing protein; ZNFD
Gene Symbols FAM170A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM170A. Recombinant protein control fragment (Product #RP-96961).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340069
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.