Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM171A2 Polyclonal Antibody

Catalog No. PIPA554077
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554077 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554077 Supplier Invitrogen™ Supplier No. PA554077

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

Immunogen sequence: RAFPAFLGTE ASSSGNGSWL ELMPLTAVSV HLLTGNGTEV PLSGPIHLSL PVPSETRALT VGTSIPAWRF DPKSGLWVRN GTGVIRKEGR QLYWTF Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.

FAM171A2 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM171A2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM171A2
Gene Accession No. A8MVW0
Gene Alias FAM171A2; family with sequence similarity 171 member A2; family with sequence similarity 171, member A2; protein FAM171A2; RGD1307966
Gene Symbols FAM171A2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM171A2. Recombinant protein control fragment (Product #RP-91006).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284069
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.