Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM171B Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300187APC
Description
FAM171B Polyclonal antibody specifically detects FAM171B in Human samples. It is validated for ELISASpecifications
FAM171B | |
Polyclonal | |
ELISA | |
family with sequence similarity 171, member B, FLJ34104, KIAA1946, NPD019 | |
Produced in rabbits immunized with purified, recombinant Human FAM171B (Accession#: AAH60872.1; Met1-Thr355) | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
165215 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction