Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM171B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552718
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552718 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552718 Supplier Invitrogen™ Supplier No. PA552718
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SKQPKHINNN LSSSLGDAQD EKRYLTGNEE AYGRSHIPEQ LMHIYSQPIA ILQTSDLFST PEQLHTAKSA TLPRKGQLVY GQLMEPVNRE NFTQTLPKMP IHSHAQPPDA REEDIILEGQ QSLPSQASDW SRYS Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 90%.

FAM171B is a protein coding gene. An important paralog of this gene is FAM171A1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM171B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM171B
Gene Accession No. Q6P995
Gene Alias D430039N05Rik; FAM171B; family with sequence similarity 171 member B; family with sequence similarity 171, member B; KIAA1946; NPD019; Protein FAM171B
Gene Symbols FAM171B
Host Species Rabbit
Immunogen Recombinant protein fragment of Human FAM171B (Product #RP-90313)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165215
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.