Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM171B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553060
Description
Immunogen sequence: FEDTVLITGK LADAKSQPSV QFSKALIKLP DNHHISNVTG YLTVLQQFLK VDNFLHTTGI TLNKPGFENI ELTPLAAICV KIYSGGKELK VNGSIQVSLP LLRLNDISAG DRIPAWTFDM NTGAWVNHGR GM Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 77%.
FAM171B is a protein coding gene. An important paralog of this gene is FAM171A1.
Specifications
| FAM171B | |
| Polyclonal | |
| Unconjugated | |
| FAM171B | |
| D430039N05Rik; FAM171B; family with sequence similarity 171 member B; family with sequence similarity 171, member B; KIAA1946; NPD019; Protein FAM171B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 165215, 241520 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14CH0, Q6P995 | |
| FAM171B | |
| Recombinant protein corresponding to Human FAM171B. Recombinant protein control fragment (Product #RP-90312). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction