Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM172A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113640
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113640 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113640 Supplier Invitrogen™ Supplier No. PA5113640
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTFKFPADPE VQSNLVGARI LREKVRARAQ GSSPRDLEGH ASSHLPSQHC For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.

FAM172A is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM172A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAM172A
Gene Accession No. Q3TNH5
Gene Alias 1110033M05Rik; 2610318O14Rik; 53-E6; 9430037D06Rik; AF064782; C5orf21; Cotranscriptional regulator FAM172A; Fam172a; family with sequence similarity 172 member A; family with sequence similarity 172, member A; LOW QUALITY PROTEIN: protein FAM172A; pEN87; Protein FAM172A; RGD1305526
Gene Symbols FAM172A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of mouse 1110033M05RIK.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68675
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.