Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM172A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546022
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546022 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546022 Supplier Invitrogen™ Supplier No. PA546022
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SSDSSDEPAE KRERKDKVSK ETKKRRDFYE KYRNPQREKE MMQLYIRENG Sequence homology: Dog: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Yeast: 83%.

FAM172A is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM172A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAM172A
Gene Accession No. Q8WUF8
Gene Alias 1110033M05Rik; 2610318O14Rik; 53-E6; 9430037D06Rik; AF064782; C5orf21; Cotranscriptional regulator FAM172A; Fam172a; family with sequence similarity 172 member A; family with sequence similarity 172, member A; LOW QUALITY PROTEIN: protein FAM172A; pEN87; Protein FAM172A; RGD1305526
Gene Symbols FAM172A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human FAM172A.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83989
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.