Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM173A Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306308PCP
Description
FAM173A Polyclonal antibody specifically detects FAM173A in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
FAM173A | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
family with sequence similarity 173, member A | |
Produced in rabbits immunized with E. coli-derived Human FAM173A fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
65990 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction