Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM173B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121095
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121095 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121095 Supplier Invitrogen™ Supplier No. PA5121095
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: A-549, BT-474, HL-60, Mouse liver, Mouse kidney, Mouse heart, Rat brain. The target is usually found in the following locations: Membrane, Single-pass membrane protein. Immunogen sequence: MEGGGGIPLE TLKEESQSRH VLPASFEVNS LQKSNWGFLL TGLVGGTLVA VYAVATPFVT PALRKVCLPF VPATTKQIEN VVKMLRCRRG SLVDIGSGDG RIVIAAAKKG FTAVGYELNP WLVWYSRYRA WREGVHGSAK FYISDLWKVT FSQYSNVVIF GVPQMMLQLE KKLERELEDD ARVIACRFPF PHWTPDHVTG EGIDTVWAYD ASTFRGREKR PCTSMHFQLP IQA.

FAM173B is a 233 amino acid single-pass membrane protein that is encoded by a gene that maps to human chromosome 5, which makes up about 6% of human genomic DNA and contains 181 million base pairs encoding around 1,000 genes. It is associated with Cockayne syndrome through the ERCC8 gene and familial adenomatous polyposis through the adenomatous polyposis coli (APC) tumor suppressor gene. Treacher Collins syndrome is also chromosome 5 associated and is caused by insertions or deletions within the TCOF1 gene. Deletion of the p arm of chromosome 5 leads to cri du chat syndrome. Deletion of 5q or chromosome 5 altogether is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM173B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FAM173B
Gene Accession No. Q6P4H8, Q9D1Z3
Gene Alias A930016P21Rik; AA987072; ATP synthase subunit C lysine N-methyltransferase; Atpsckmt; FAM173B; family with sequence similarity 173 member B; family with sequence similarity 173, member B; hFAM173B; JS-2; mFam173b; protein FAM173B; Protein N-lysine methyltransferase FAM173B; RGD1560629
Gene Symbols FAM173B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-233 of human FAM173B (NP_954584.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134145, 499561, 68073
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.