Learn More
Invitrogen™ FAM174A Monoclonal Antibody (1F3)

Description
Peptide Sequence: MKASQCCCCL SHLLASVLLL LLLPELSGPL AVLLQAAEAA PGLGPPDPRP RTLPPLPPGP TPAQQPGRGL AEAAGPRGSE GGNGSNPVAG LETDDHGGKA GEGSVGGGLA VSPNPGDKPM TQRALTVLMV VSGAVLVYFV VRTVRMRRRN RKTRRYGVLD TNIENMELTP LEQDDEDDDN TLFDANHPRR.
Specifications
Specifications
| Antigen | FAM174A |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1F3 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | FAM174A |
| Gene Accession No. | Q8TBP5 |
| Gene Alias | FAM174A; family with sequence similarity 174 member A; family with sequence similarity 174, member A; HCV NS5A-transactivated protein 6; Hepatitis C virus NS5A-transactivated protein 6; HGS_RE408; Membrane protein FAM174A; NS5ATP6; TMEM157; Transmembrane protein 157; UNQ1912; UNQ1912/PRO4371 |
| Gene Symbols | FAM174A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.