Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM174A Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 248021AP150UL

Specifications
FAM174A | |
Polyclonal | |
Unconjugated | |
FAM174A | |
FAM174A, Membrane protein FAM174A, NS5ATP6, TMEM157, Transmembrane protein 157, UNQ1912 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
345757 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8TBP5 | |
FAM174A | |
FAM174A Fusion Protein Ag20431 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction