Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM174B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542240
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542240 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542240 Supplier Invitrogen™ Supplier No. PA542240
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MRAVPLPAPL LPLLLLALLA APAARASRAE SVSAPWPEPE RESRPPPGPG Sequence homology: Human: 93%.

Essential for Golgi structural integrity. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM174B
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAM174B
Gene Accession No. Q3ZCQ3
Gene Alias fam174b; family with sequence similarity 174 member B; family with sequence similarity 174, member B; membrane protein FAM174B; si:ch211-196l7.3; Unknown (protein for MGC:140090)
Gene Symbols FAM174B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human LOC400451 (aa 1-50).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 400451
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.