Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM175B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501735482 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5482 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5482 Supplier Proteintech Group Inc Supplier No. 268681AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Component of the BRISC complex, a multiprotein complex that specifically cleaves ′Lys-63′-linked polyubiquitin, leaving the last ubiquitin chain attached to its substrates. May act as a central scaffold protein that assembles the various components of the BRISC complex and retains them in the cytoplasm. Plays a role in regulating the onset of apoptosis via its role in modulating ′Lys-63′-linked ubiquitination of target proteins. Required for normal mitotic spindle assembly and microtubule attachment to kinetochores via its role in deubiquitinating NUMA1. Plays a role in interferon signaling via its role in the deubiquitination of the interferon receptor IFNAR1; deubiquitination increases IFNAR1 activities by enhancing its stability and cell surface expression. Down-regulates the response to bacterial lipopolysaccharide (LPS) via its role in IFNAR1 deubiquitination. Required for normal induction of p53/TP53 in response to DNA damage. Independent of the BRISC complex, promotes interaction between USP7 and p53/TP53, and thereby promotes deubiquitination of p53/TP53, preventing its degradation and resulting in increased p53/TP53-mediated transcription regulation and p53/TP53-dependent apoptosis in response to DNA damage.

Specifications

Antigen FAM175B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ABRAXAS2
Gene Accession No. Q15018, Q3TCJ1
Gene Alias Abraxas brother protein 1, ABRO1, BRISC complex subunit Abro1, FAM175B, KIAA0157, Protein FAM175B
Gene Symbols ABRAXAS2
Host Species Rabbit
Immunogen FAM175B Fusion Protein Ag22121
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109359, 23172
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.