Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM177A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562762
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562762 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562762 Supplier Invitrogen™ Supplier No. PA562762
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NRMSEEAEKQ YQQNKLQTDS IVQTDQPETV ISSSFVNVNF EMEGDSEVIM ESKQNPVSVP Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 80%.

This gene encodes a member of a conserved protein family. Alternative splicing results in multiple transcript variants. This gene is thought to be associated with susceptibility to juvenile idiopathic arthritis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM177A1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM177A1
Gene Accession No. Q8N128
Gene Alias C14orf24; FAM177A1; family with sequence similarity 177 member A1; family with sequence similarity 177, member A1; protein FAM177A1
Gene Symbols FAM177A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM177A1. Recombinant protein control fragment (Product #RP-101372).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283635
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.