Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM177B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564088
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564088 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564088 Supplier Invitrogen™ Supplier No. PA564088
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TQPKYQYVLN EFYRIQNKKS DNKSERRGSK AQAAEVPNEK CHLEAGVREY GTIQQDVTEA IPQ Highest antigen sequence identity to the following orthologs: Mouse - 28%, Rat - 48%.

FAM177B is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM177B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM177B
Gene Accession No. A6PVY3
Gene Alias FAM177B; family with sequence similarity 177 member B; family with sequence similarity 177, member B; protein FAM177B
Gene Symbols FAM177B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM177B. Recombinant protein control fragment (Product #RP-103489).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 400823
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.