Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM178A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566091
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566091 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566091 Supplier Invitrogen™ Supplier No. PA566091
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASKYLAKGTNI YVPSSYHLPK EMKSLKKKHR SPERRKSLFI HENNEKNDRD RGKTNADSKK QTTVAEADIF NNSSRSLS Highest antigen sequence identity to the following orthologs - mouse 71%, rat 71%.

Plays a role in the DNA damage response (DDR) pathway by regulating postreplication repair of UV-damaged DNA and genomic stability maintenance (PubMed:25931565). The SLF1-SLF2 complex acts to link RAD18 with the SMC5-SMC6 complex at replication-coupled interstrand cross-links (ICL) and DNA double-strand breaks (DSBs) sites on chromatin during DNA repair in response to stalled replication forks (PubMed:25931565). Promotes the recruitment of the SMC5-SMC6 complex to DNA lesions (PubMed:25931565).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM178A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLF2
Gene Accession No. Q8IX21
Gene Alias C10orf6; FAM178A; family with sequence similarity 178, member A; protein FAM178A; SLF2; Smc5/6 localization factor 1; SMC5-SMC6 complex localization factor 2; SMC5-SMC6 complex localization factor protein 2
Gene Symbols SLF2
Host Species Rabbit
Immunogen Recombinant Human FAM178A. Recombinant protein control fragment (Product #RP-105077).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55719
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.