Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM179B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565645
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565645 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565645 Supplier Invitrogen™ Supplier No. PA565645
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DTDWLLAGNRT QSAHCHCGDH VRDSMHIYGS YSPTICTRRV LSAGKGKNKL PWENEQPGIM GENQTSTSKD IEQFSTYDFI PSAKLKLSQG MPVNDDLCFS Highest antigen sequence identity to the following orthologs - mouse 83%, rat 85%.

Required for normal structure and function of primary cilia. Plays a role in the organization of axoneme microtubule bundles in primary cilia. Interacts with microtubules and promotes microtubule polymerization via its HEAT repeat domains, especially those in TOG region 2 and 4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM179B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TOGARAM1
Gene Accession No. Q9Y4F4
Gene Alias Crescerin-1; FAM179B; family with sequence similarity 179 member B; family with sequence similarity 179, member B; KIAA0423; Protein FAM179B; TOG array regulator of axonemal microtubules protein 1; TOGARAM1
Gene Symbols TOGARAM1
Host Species Rabbit
Immunogen Recombinant Human FAM179B. Recombinant protein control fragment (Product #RP-106317).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23116
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.