Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM180B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556133
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556133 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556133 Supplier Invitrogen™ Supplier No. PA556133
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PEVMFELLWA GLELDVMGQL HIQDEELAST HPGRRLRLLL QHHVPSDLEG TEQWLQQLQD LRKGPPLSTW DFEHLLLTGL SCVYRLHAAS EAEERGRWTQ VFALLAQETL WDLCKGFCP Highest antigen sequence identity to the following orthologs: Mouse - 28%, Rat - 28%.

FAM180B (Family With Sequence Similarity 180 Member B) is a Protein Coding gene. Diseases associated with FAM180B include Leprosy 3. An important paralog of this gene is FAM180A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM180B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM180B
Gene Accession No. Q6P0A1
Gene Alias FAM180B; family with sequence similarity 180 member B; family with sequence similarity 180, member B; hypothetical gene supported by BC065704; protein FAM180B
Gene Symbols FAM180B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM180B. Recombinant protein control fragment (Product #RP-90587).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 399888
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.