Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM181B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571108
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571108 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571108 Supplier Invitrogen™ Supplier No. PA571108
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%; Zebrafish: 93%.

FAM181B is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM181B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAM181B
Gene Accession No. A6NEQ2
Gene Alias A830059I20Rik; AB041569; FAM181B; family with sequence similarity 181 member B; family with sequence similarity 181, member B; LOW QUALITY PROTEIN: protein FAM181B; protein FAM181B; RGD1564695
Gene Symbols FAM181B
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human FAM181B.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220382
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.