Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM182A Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144139
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144139 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144139 Supplier Invitrogen™ Supplier No. PA5144139
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM182A Polyclonal Antibody for ICC/IF, ELISA

FAM182A is a gene that encodes a protein called Family with sequence similarity 182 member A. The specific function of FAM182A is not yet fully understood, and further research is required to elucidate its role in cellular processes and its potential implications for human health and disease. It is worth noting that FAM182A has been found to be expressed in various tissues and cell types, and alterations in its expression have been associated with certain diseases, including cancer. Therefore, understanding the specific functions and mechanisms of FAM182A may provide valuable insights into its potential role in disease development and progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM182A
Applications ELISA, Immunocytochemistry
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene FAM182A
Gene Accession No. Q5T1J6
Gene Alias C20orf91
Gene Symbols FAM182A
Host Species Rabbit
Immunogen Recombinant Human Protein FAM182A protein (1-154AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284800
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.