Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM183B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571109
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571109 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571109 Supplier Invitrogen™ Supplier No. PA571109
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 92%; Guinea Pig: 79%; Horse: 100%; Human: 100%; Mouse: 79%; Pig: 93%; Rabbit: 92%; Rat: 79%.

FAM183BP is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM183B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAM183BP
Gene Accession No. Q6ZVS7
Gene Alias 3100002J23Rik; acyloxyacyl hydrolase (neutrophil); FAM183B; FAM183BP; family with sequence similarity 183, member B; protein FAM183B; RGD1561932; testis highly expressed protein 6; THEG6
Gene Symbols FAM183BP
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human FAM183B.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340286
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.