Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM184A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556977
Description
Immunogen sequence: ELDTLKRSQL FTAESLQASK EKEADLRKEF QGQEAILRKT IGKLKTELQM VQDEAGSLLD KCQKLQTALA IAENNVQVLQ KQLDDAKEGE MALLSKHKEV GSEL Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
FAM184A (Family With Sequence Similarity 184 Member A) is a Protein Coding gene. An important paralog of this gene isFAM184B.
Specifications
FAM184A | |
Polyclonal | |
Unconjugated | |
FAM184A | |
3110012E06Rik; 4930438C08Rik; 4930589M24Rik; C6orf60; FAM184A; Fam184a-ps1; family with sequence similarity 184 member A; family with sequence similarity 184, member A; family with sequence similarity 184, member A, pseudogene 1; protein FAM184A; RGD1311294 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79632 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NB25 | |
FAM184A | |
Recombinant protein corresponding to Human FAM184A. Recombinant protein control fragment (Product #RP-94650). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction