Learn More
Invitrogen™ FAM184A Polyclonal Antibody

Description
Immunogen sequence: KVNEAKRTQQE YYERELKNLQ SRLEEEVTQL NEAHSKTLEE LAWKHHMAIE AVHSNAIRDK KKLQMDLEEQ HNKDKLNLE Highest antigen sequence identity to the following orthologs - mouse 83%, rat 83%.
Specifications
Specifications
| Antigen | FAM184A |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | FAM184A |
| Gene Accession No. | Q8NB25 |
| Gene Alias | 3110012E06Rik; 4930438C08Rik; 4930589M24Rik; C6orf60; FAM184A; Fam184a-ps1; family with sequence similarity 184 member A; family with sequence similarity 184, member A; family with sequence similarity 184, member A, pseudogene 1; protein FAM184A; RGD1311294 |
| Gene Symbols | FAM184A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.