Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM184B Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300242JF646
Description
FAM184B Polyclonal antibody specifically detects FAM184B in Human samples. It is validated for Western BlotSpecifications
FAM184B | |
Polyclonal | |
Western Blot | |
family with sequence similarity 184, member B, hypothetical protein LOC27146, KIAA1276 | |
A synthetic peptide mapping to a region between residue 1000 and the C-terminus (residue 1068) of human KIAA1276 using the numbering given in TrEMBL entry Q9ULE4 (GeneID 27146). | |
0.1 mL | |
Neuroscience | |
27146 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction