Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM187B Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144133
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144133 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144133 Supplier Invitrogen™ Supplier No. PA5144133
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM187B Polyclonal Antibody for IHC (P), ELISA

FAM187B is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM187B
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene FAM187B
Gene Accession No. Q17R55
Gene Alias FAM187B; family with sequence similarity 187 member B; family with sequence similarity 187, member B; protein FAM187B; TMEM162; transmembrane protein 162
Gene Symbols FAM187B
Host Species Rabbit
Immunogen Recombinant Human Protein FAM187B protein (18-335AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148109
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.