Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM189A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552717
Description
Immunogen sequence: MVSSDVLQMF LPQRSHPANP TCVTPHGTVL HQTLDFDEFI PPLPPPPYYP PEYTCTPSTE AQRGLHLDFA PSPFGTLYDV AINSPGLLYP AELPPPYEAV VGQPPASQVT SIGQQVAESS SGDPNTSAGF STPVP Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 84%.
Specifications
| FAM189A1 | |
| Polyclonal | |
| Unconjugated | |
| FAM189A1 | |
| FAM189A1; family with sequence similarity 189 member A1; family with sequence similarity 189, member A1; KIAA0574; Protein FAM189A1; TMEM228; Transmembrane protein 228 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23359 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O60320 | |
| FAM189A1 | |
| Recombinant protein corresponding to Human FAM189A1. Recombinant protein control fragment (Product #RP-90040). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction