Learn More
Invitrogen™ FAM189A2 Polyclonal Antibody

Description
Immunogen sequence: MSEGSEAAVI PLDLGCTQVT QDGDIPNIPA EENASTSTPS STLVRPIRSR RALPPLRTRS KSDPVLHPSE ER Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 64%.
Specifications
Specifications
| Antigen | FAM189A2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | FAM189A2 |
| Gene Accession No. | Q15884 |
| Gene Alias | AI414343; AI666772; C9orf61; Fam189a2; family with sequence similarity 189 member A2; family with sequence similarity 189, member A2; Friedreich ataxia region gene X123; Gm967; Protein FAM189A2; Protein X123; X123 |
| Gene Symbols | FAM189A2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.