Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM189A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556135
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556135 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556135 Supplier Invitrogen™ Supplier No. PA556135
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PVLSCEAATQ TERRLDLAAV TLRRGLRSRA SRCRPRSLID YKSYMDTKLL VARFLEQSSC TMTPDIHELV ENIKSVLKSD EEHMEEAITS ASFL Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.

C9orf61, also named as FAM189A2 and X123, is predominantly expressed in muscle. It has three isoforms with MW 45-50 kDa, 32-35 kDa and 27 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM189A2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM189A2
Gene Accession No. Q15884
Gene Alias AI414343; AI666772; C9orf61; Fam189a2; family with sequence similarity 189 member A2; family with sequence similarity 189, member A2; Friedreich ataxia region gene X123; Gm967; Protein FAM189A2; Protein X123; X123
Gene Symbols FAM189A2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM189A2. Recombinant protein control fragment (Product #RP-90961).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9413
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.