Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM18B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554030
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554030 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554030 Supplier Invitrogen™ Supplier No. PA554030
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RCKVRSRKHL TSMATSYFGK QFLRQNTGDD QTS Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 82%.

The FAM18B gene (Family with Sequence Similarity 18 Member B) is located on chromosome 17q21.31 and encodes a protein whose precise biological functions are not fully characterized. The FAM18 family, to which FAM18B belongs, includes proteins that are presumed to be involved in cellular processes such as signal transduction and intracellular trafficking. Although the specific role of FAM18B remains under investigation, initial studies suggest it may be involved in modulating cellular responses to environmental stimuli and maintaining cellular homeostasis. There is limited research on FAM18B's involvement in disease, but its expression patterns in various tissues, including the brain and reproductive organs, indicate potential roles in neurological and reproductive functions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM18B
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TVP23B
Gene Accession No. Q9NYZ1
Gene Alias CGI-148; FAM18B; FAM18B1; family with sequence similarity 18, member B; family with sequence similarity 18, member B1; Golgi apparatus membrane protein TVP23 homolog B; NPD008; protein FAM18B1; trans-golgi network vesicle protein 23 homolog B (S. cerevisiae); TVP23B; YDR084C
Gene Symbols TVP23B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM18B. Recombinant protein control fragment (Product #RP-91108).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51030
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.