Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM190A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | FAM190A |
---|---|
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
FAM190A Polyclonal specifically detects FAM190A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FAM190A | |
Polyclonal | |
Rabbit | |
Human | |
family with sequence similarity 190, member A, KIAA1680 protein, KIAA1680MGC48628 | |
CCSER1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
401145 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:KEHSTNKNVFINCLSSGKSEGDDSGFTEDQTRRSVKQSTRKLLPKSFSSHYKFSKPVLQSQSISLVQQSEFSLEVTQYQEREPVLVRASPSCSVDVTERA | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title