Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM193A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571111
Description
This target displays homology in the following species: Dog: 85%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 93%; Rat: 93%; Zebrafish: 85%.
FAM193A is a protein coding gene.
Specifications
FAM193A | |
Polyclonal | |
Unconjugated | |
FAM193A | |
C4orf8; Fam193a; family with sequence similarity 193 member A; family with sequence similarity 193, member A; im:7155132; Protein FAM193A; Protein IT14; RES4-22; RGD1309634 | |
Rabbit | |
Affinity chromatography | |
RUO | |
8603 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P78312 | |
FAM193A | |
synthetic peptide directed towards the C-terminal region of human FAM193A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction