Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM193B Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281932
Description
FAM193B Polyclonal specifically detects FAM193B in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
FAM193B | |
Polyclonal | |
Unconjugated | |
PBS | |
FAM193B | |
FAM193B antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human FAM193B. The immunogen is located within amino acids 120 - 170 of FAM193B. Amino Acid Squence: RKSMEEDERQTGREHA | |
Affinity Purified | |
RUO | |
54540 | |
Human | |
IgG |
Western Blot, ELISA, Immunofluorescence | |
1 mg/mL | |
Western Blot 1 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 20 μg/mL | |
DKFZp313I142, family with sequence similarity 193, member B, KIAA1931 | |
Rabbit | |
99 kDa | |
0.1 mg | |
Primary | |
At least two isoforms of FAM193B are known to exist; this antibody will detect both isoforms. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction