Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM193B Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281932MFV610
Description
FAM193B Polyclonal antibody specifically detects FAM193B in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| FAM193B | |
| Polyclonal | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| DKFZp313I142, family with sequence similarity 193, member B, KIAA1931 | |
| FAM193B antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human FAM193B. The immunogen is located within amino acids 120 - 170 of FAM193B. Amino Acid Squence: RKSMEEDERQTGREHA | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 54540 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction