Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM193B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557427
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557427 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557427 Supplier Invitrogen™ Supplier No. PA557427
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NSTEPKVPNS ARAAKRARHK LKKKEKEKAQ LAAEALKQAN RVSGSREPRP ARERLLEWPD RELDRVNSFL SSRLQEIKNT VKDSIRASFS VCE Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.

FAM193B, also known as IRIZIO, was initially identified as a protein that is upregulated in alveolar rhabdomyosarcoma (ARMS), a type of fast-growing tumor characterized by chromosomal translocations fusing the PAX3 or PAX7 gene with that of FOXO1. It has been suggested that, in addition to the PAX3-FOXO1 gene fusion, disruption of the Rb and p53 pathways is required for full ARMS tumorigenesis. In PAX3-FOXO1-expressing primary mouse fibroblasts that possessed a defective p53 pathway, FAM193B expression enabled the tumorigenic transformation, suggesting that FAM193B may contribute to rhabdomyosarcomagenesis in humans.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM193B
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM193B
Gene Accession No. Q96PV7
Gene Alias FAM193B; family with sequence similarity 193 member B; IRIZIO; KIAA1931; Protein FAM193B
Gene Symbols FAM193B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM193B. Recombinant protein control fragment (Product #RP-94989).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54540
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.