Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM193B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140060
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: NTVPNLSRVI WVKTPKPGYP SSEEPSSKEV PSCKQELPEP VSSGGKPQKG KRQGSQAKKS EASPAPRPPA SLEVPSAKGQ VAGPKQ.
FAM193B, also known as IRIZIO, was initially identified as a protein that is upregulated in alveolar rhabdomyosarcoma (ARMS), a type of fast-growing tumor characterized by chromosomal translocations fusing the PAX3 or PAX7 gene with that of FOXO1. It has been suggested that, in addition to the PAX3-FOXO1 gene fusion, disruption of the Rb and p53 pathways is required for full ARMS tumorigenesis. In PAX3-FOXO1-expressing primary mouse fibroblasts that possessed a defective p53 pathway, FAM193B expression enabled the tumorigenic transformation, suggesting that FAM193B may contribute to rhabdomyosarcomagenesis in humans.
Specifications
| FAM193B | |
| Polyclonal | |
| Unconjugated | |
| FAM193B | |
| FAM193B; family with sequence similarity 193 member B; IRIZIO; KIAA1931; Protein FAM193B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54540 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96PV7 | |
| FAM193B | |
| Recombinant protein corresponding to Human FAM193B. Recombinant protein control fragment (Product #RP-106816). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction