Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM195A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104344
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104344 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104344 Supplier Invitrogen™ Supplier No. PA5104344
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total C16orf14.

FAM195A gene ontology (GO) annotation includes protein binding and nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM195A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MCRIP2
Gene Accession No. Q9BUT9, Q9CQB2
Gene Alias 9530058B02Rik; C16orf14; c349E10.1; FAM195A; family with sequence similarity 195 member A; family with sequence similarity 195, member A; MAPK regulated corepressor interacting protein 2; MAPK regulated co-repressor interacting protein 2; MCRIP2; Protein FAM195A
Gene Symbols MCRIP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human MCRIP2(Accession Q9BUT9), corresponding to amino acid residues R15-R65.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68241, 685545, 84331
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.