Learn More
Invitrogen™ FAM195A Polyclonal Antibody

Description
Immunogen sequence: EESVRFVSEA WQQVQQQLDG GPAGEGGPRP VQYVERTPNP RLQNFVPIDL DEWWAQQFLA RITSC Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 85%.
Specifications
Specifications
| Antigen | FAM195A |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MCRIP2 |
| Gene Accession No. | Q9BUT9 |
| Gene Alias | 9530058B02Rik; C16orf14; c349E10.1; FAM195A; family with sequence similarity 195 member A; family with sequence similarity 195, member A; MAPK regulated corepressor interacting protein 2; MAPK regulated co-repressor interacting protein 2; MCRIP2; Protein FAM195A |
| Gene Symbols | MCRIP2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.