Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM196B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561471
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561471 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561471 Supplier Invitrogen™ Supplier No. PA561471
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AEVDVQTPED PAVMGKTQAT RHHLPPTYSL SFPRSQKAGG FRNIAIQTSP SLRKHFPVFK RKRLTASKSL VEMPTASQSA IQVNGN Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 87%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM196B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM196B
Gene Accession No. A6NMK8
Gene Alias C5orf57; FAM196B; family with sequence similarity 196 member B; family with sequence similarity 196, member B; Inhibitory synaptic factor family member 2B; INSYN2B; protein FAM196B; Protein INSYN2B
Gene Symbols FAM196B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM196B. Recombinant protein control fragment (Product #RP-100312).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100131897
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.