Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM198B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116209
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116209 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116209 Supplier Invitrogen™ Supplier No. PA5116209
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM198B Polyclonal Antibody for Western Blot

FAM198B is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM198B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene FAM198B
Gene Accession No. Q6UWH4
Gene Alias 1110032E23Rik; 2210419I08Rik; AD021; AD036; AV011458; C4orf18; Ened; Expressed in nerve and epithelium during development; FAM198B; family with sequence similarity 198 member B; family with sequence similarity 198, member B; gask1b; GaskbB; golgi associated kinase 1B; Golgi-associated kinase 1B; LOW QUALITY PROTEIN: protein FAM198B; protein ENED; protein ENED; protein FAM198B; protein FAM198B; UNQ2512/PRO6001
Gene Symbols FAM198B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 275-304 amino acids from the Central region of human C4orf18.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51313
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.