Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM199X Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551823
Description
Immunogen sequence: WSAMTNDEQV EYIEYLSRKV STEMGLREQL DIIKIIDPSA QISPTDSEFI IELNCLTDEK LKQVRNYIKE HSPRQRPARE AWKRSNFSCA STSGVSGASA SASSSSASMV SSASSSGSSV GNSASNSSA Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 62%.
Specifications
| FAM199X | |
| Polyclonal | |
| Unconjugated | |
| FAM199X | |
| CXorf39; FAM199X; family with sequence similarity 199, X-linked; protein FAM199X | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 139231 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6PEV8 | |
| FAM199X | |
| Recombinant protein corresponding to Human FAM199X. Recombinant protein control fragment (Product #RP-92196). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction