Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM19A3 Polyclonal Antibody
Rabbit Polyclonal Antibody
$237.00 - $539.50
Specifications
Antigen | FAM19A3 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119103
|
Invitrogen™
PA5119103 |
200 μL |
Each for $539.50
|
|
|||||
PIPA5122004
|
Invitrogen™
PA5122004 |
20 μL |
Each for $237.00
|
|
|||||
Description
FAM19A3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
FAM19A3 | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
FAM19A3 | |
Chemokine-like protein TAFA-3; FAM19A3; family with sequence similarity 19 (chemokine (C-C motif)-like), member A3; family with sequence similarity 19 member A3, C-C motif chemokine like; protein FAM19A3; TAFA3; TAFA-3 | |
FAM19A3 | |
Primary | |
Antigen Affinity Chromatography |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q7Z5A8 | |
284467 | |
Recombinant protein Family With Sequence Similarity 19, Member A3. The antigen corresponds to amino acid range 44-130 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title