Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM200A Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305968H
Description
FAM200A Polyclonal specifically detects FAM200A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FAM200A | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C7orf38, chromosome 7 open reading frame 38, DKFZp727G131, family with sequence similarity 200, member A, FLJ36794, hypothetical protein LOC221786 | |
Produced in rabbits immunized with E. coli-derived Human FAM200A fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HRP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
221786 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction