Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM204A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566447
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566447 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566447 Supplier Invitrogen™ Supplier No. PA566447
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QWKELTQYFGV NDRFDPPVKR KKVEKSGLEK RIDQAVEEWN IEKAEELSNQ L Highest antigen sequence identity to the following orthologs - mouse 88%, rat 88%.

FAM204A is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM204A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM204A
Gene Accession No. Q9H8W3
Gene Alias bA319I23.1; C10orf84; FAM204A; family with sequence similarity 204 member A; family with sequence similarity 204, member A; protein FAM204A
Gene Symbols FAM204A
Host Species Rabbit
Immunogen Recombinant Human FAM204A. Recombinant protein control fragment (Product #RP-107122).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63877
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.