Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM204A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546216
Description
Peptide sequence: NSGLNLQEDK EDESIRKTEI IDFSTDEPKT ETESNVNAYE ECPSGIPIDM Sequence homology: Dog: 79%; Horse: 79%; Human: 100%.
FAM204A is a protein coding gene.
Specifications
| FAM204A | |
| Polyclonal | |
| Unconjugated | |
| FAM204A | |
| bA319I23.1; C10orf84; FAM204A; family with sequence similarity 204 member A; family with sequence similarity 204, member A; protein FAM204A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 63877 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H8W3 | |
| FAM204A | |
| Synthetic peptide directed towards the N-terminal of human FAM204A (aa 27-76). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction