Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM206A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557460
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557460 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557460 Supplier Invitrogen™ Supplier No. PA557460
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ATEPEAAEPV VPSLVDRYFT RWYKPDVKGK FCEDHCILQH SNRICVITLA ESHPVLQSGK TIKSISYQIS TNCSRLQNKV S Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 84%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM206A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM206A
Gene Accession No. Q9NX38
Gene Alias ABITRAM; Actin-binding transcription modulator; C9orf6; CG-8; FAM206A; family with sequence similarity 206 member A; family with sequence similarity 206, member A; Protein Abitram; protein FAM206A; protein Simiate; simiate
Gene Symbols FAM206A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM206A. Recombinant protein control fragment (Product #RP-96556).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54942
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.