Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM208A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589059
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589059 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589059 Supplier Invitrogen™ Supplier No. PA589059
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DTEDMRSKTV LKRKLEDLPE NMRKLAKTSN LSENCHLYEE SPQPIGSLGH DADLRRQQQD TCNSGIADIH RLFNWLSETL ANARHSDASL TDTVNKALGL STDDAYEELR QKHEYELNST PDKKDYEQPT CAKVENAQFK GTQSLLLEVD ATSKYSVAIS TSEVGTDHKL HLKEDPNLIS VNNFEDCSLC PSVPIEHGFR RQQSKSNNVE ETEIHWKLIP ITGGNARSPE DQLGKHGEKQ TPGMKSPEEQ LVCVPPQEAF PNDPRVINRQ RSSDYQFPSS PFTDTLKGTT EDDVLTGQVE E; Positive Samples: U-87MG, HeLa, Jurkat, HepG2; Cellular Location: Chromosome, Nucleus.

TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM208A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene TASOR
Gene Accession No. Q9UK61
Gene Alias C3orf63; CTCL tumor antigen se89-1; FAM208A; family with sequence similarity 208 member A; family with sequence similarity 208, member A; KIAA1105; protein FAM208A; protein TASOR; RAP140; retinoblastoma-associated protein 140; retinoblastoma-associated protein RAP140; se89-1; TASOR; Transgene activation suppressor protein
Gene Symbols TASOR
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23272
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.