Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM208A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552335
Description
Immunogen sequence: TIIIKASAKG GNLPPVSPND SGAKIASNPL ERHVIPVSSS DFNNKHLLEP LCSDPLKDTN SDEQHSTSAL TEVEMNQPQH ATELMVTSDH IVPGDMAREP VEETTKSPSD VNISAQPALS N Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 66%.

Specifications
FAM208A | |
Polyclonal | |
Unconjugated | |
TASOR | |
C3orf63; CTCL tumor antigen se89-1; FAM208A; family with sequence similarity 208 member A; family with sequence similarity 208, member A; KIAA1105; protein FAM208A; protein TASOR; RAP140; retinoblastoma-associated protein 140; retinoblastoma-associated protein RAP140; se89-1; TASOR; Transgene activation suppressor protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23272 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UK61 | |
TASOR | |
Recombinant protein corresponding to Human FAM208A. Recombinant protein control fragment (Product #RP-89490). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction