Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ FAM209A Overexpression Lysate
SDP

Catalog No. NBL108355 Shop All R&D Systems Products
Catalog No. NBL108355 Supplier Novus Biologicals™ Supplier No. NBL108355
Add to Cart
Add to Cart
The C20orf106 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.

Specifications

Cell Line The lysate was created in HEK293T cells, using plasmid ID RC21459 and based on accession number NM_112971. The protein contains a C-terminal DDK tag.
For Use With (Application) Western Blot
Storage Buffer RIPA buffer
Purity Protein
Test Specificity Homo sapiens chromosome 20 open reading frame 106 (C20orf106), mRNA.
Content And Storage -80°C. Avoid freeze-thaw cycles.
Target FAM209A
Protein State Native

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.