Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM20A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561848
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561848 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561848 Supplier Invitrogen™ Supplier No. PA561848
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RHFPTISADY SQDEKALLGA CDCTQIVKPS GVHLKLVLRF SDFGKAMFKP MRQQRDEETP VDFFYFIDFQ RH Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 94%.

This locus encodes a protein that is likely secreted and may function in hematopoiesis. A mutation at this locus has been associated with amelogenesis imperfecta and gingival hyperplasia syndrome. Alternatively spliced transcript variants have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM20A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM20A
Gene Accession No. Q96MK3
Gene Alias AI1G; AI606893; AIGFS; FAM20A; family with sequence similarity 20 member A; family with sequence similarity 20, member A; FP2747; ortholog of human family with sequence similarity 20, member A FAM20A; protein FAM20A; pseudokinase FAM20A; RGD1306364; UNQ9388/PRO34279
Gene Symbols FAM20A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM20A. Recombinant protein control fragment (Product #RP-100911).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54757
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.