Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM20A Polyclonal Antibody, Invitrogen™

Catalog No. PIPA524891
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524891 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524891 Supplier Thermo Scientific Supplier No. PA524891

Rabbit Polyclonal Antibody

This locus encodes a protein that is likely secreted and may function in hematopoiesis. A mutation at this locus has been associated with amelogenesis imperfecta and gingival hyperplasia syndrome. Alternatively spliced transcript variants have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM20A
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene FAM20A
Gene Accession No. Q96MK3
Gene Alias AI1G; AI606893; AIGFS; FAM20A; family with sequence similarity 20 member A; family with sequence similarity 20, member A; FP2747; ortholog of human family with sequence similarity 20, member A FAM20A; protein FAM20A; pseudokinase FAM20A; RGD1306364; UNQ9388/PRO34279
Gene Symbols FAM20A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 48-77 amino acids from the N-terminal region of human FA20A
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54757
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.